Safety ankle boots, lace-up, suXXeed® S3, Heckel
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6390335 6390336 6390337 6390338 6390339 6390342 6390343 6390344 6390340 6390341 6390347 6390345 6390346
UVEX6390335EA 0 EUR
UVEX6390335 UVEX6390336 UVEX6390337 UVEX6390338 UVEX6390339 UVEX6390342 UVEX6390343 UVEX6390344 UVEX6390340 UVEX6390341 UVEX6390347 UVEX6390345 UVEX6390346
Safety ankle boots, lace-up, suXXeed® S3, Heckel
Schoenen en laarzen
Metal-free, flexible and lightweight safety ankle boot made from oiled hydro suede leather upper resistant to liquid penetration, collar and gusset in water-repellent fabric, fabric eyelets and a double desity PU2D sole, highly slip resistant and resistant to oils and hydrocarbons.
  • Quick drying Climate Airmesh lining, very resistant to abrasion, very good absoprtion of water vapour
  • Removable insole, top in abrasion-resistant fabric
  • Metal-free protective toecap (200 J)
  • Metal-free puncture resistant midsole
Suitable for heavy industry, warehousing and logistics.
Colour: Black/red
Certificaten: EN ISO 20345:2011; S3, CI, SRC