Diaphragm pumps, VARIO® select
Leverancier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20736350 20738150
131-0033EA 5800 EUR
131-0033 131-0050
Diaphragm pumps, VARIO® select
Pompen Membraanvacuümpompen
The VACUU·SELECT controller simplifies your work and allows VARIO select diaphragm pumps to deliver unparalleled precision. The pumps run whisper quiet and respond to demand in real time by matching pumping speed to process conditions. The intuitive VACUU·SELECT controller includes predefined applications for all common vacuum processes. VACUUBRAND VARIO® technology - frees you up for other tasks.
  • VACUU·SELECT vacuum controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Contamination-free pumping and evacuation
  • Extraordinary diaphragm life for minimum operational and servicing costs
  • Excellent ultimate vacuum and TURBO·MODE for use as backing pump for turbo pumps
All wetted parts are made of aluminium, stainless steel and carefully selected plastics suitable for a wide range of applications with non-corrosive gases.
MD 4 VARIO® select
This three-stage diaphragm pump is an excellent solution for continuous, oil-free pumping of non-corrosive gases to a vacuum level as deep as 1 mbar.
MV 2 VARIO® select
Four-stage diaphragm pumps - like the MV 2 VARIO select - are an excellent solution for continuous, oil-free pumping of non-corrosive gases at fractional millibar vacuum levels.
MD 12 VARIO® select
This three-stage diaphragm pump is a powerful solution for continuous, oil-free pumping of non-corrosive gases to a vacuum level as deep as 1,5 mbar. With its high pumping speed, this pump is an excellent option for higher gas load applications or rapid pump down of vacuum chambers.
MV 10 VARIO® select
Four-stage diaphragm pumps like the MV 10 VARIO select are excellent solutions for continuous, oil-free pumping of non-corrosive gases to vacuum levels that are only a fraction of a mbar. With its high pumping speed, this pump is an excellent option for higher gas load applications or rapid pump down of vacuum chambers.
Bestelinformatie: Pump completely mounted, ready for use, with operating manual.