Industrial gloves, uvex compact NB27E
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6094609 6094610
UVEX6094609EA 53.8 EUR
UVEX6094609 UVEX6094610
Industrial gloves, uvex compact NB27E
Handschoenen
These robust cotton jersey gloves feature a special NBR coating on palm, fingers and ³/₄ of the back of the hand. They are ideal for rough work and handling coarse materials.
  • Very good mechanical abrasion resistance
  • High tear resistance
Areas of application: Iron and steel industry, shipping, logistics, concrete and construction work, wood processing.
Certificaten: EN 388:2016 (4121X)