Heating tapes, IT-H
Leverancier: ISOPAD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
802236-000 433904-000 965602-000 444118-000 740644-000
ISOP802236-000EA 426.1 EUR
ISOP802236-000 ISOP433904-000 ISOP965602-000 ISOP444118-000 ISOP740644-000
Heating tapes, IT-H
Verwarmers
Isopad IT-H heating tape is suitable where high power input is needed or where work has to be carried out at high temperatures.
  • Working element temperature of up to 900 °C
  • Non-hazardous, ordinary area
  • Storage temperature –40 to +50 °C
This tape should only be used in dry atmospheres with additional electrical protection and always with a temperature regulator.