NMR tubes, polyethylene pressure caps
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP897095-0001 SP897095-0021 SP897095-0061 SP897095-0071 SP897095-0081 SP897095-1001
SCERSP897095-0001EA 26.4 EUR
SCERSP897095-0001 SCERSP897095-0021 SCERSP897095-0061 SCERSP897095-0071 SCERSP897095-0081 SCERSP897095-1001
NMR tubes, polyethylene pressure caps
Sluitingen Reageerbuissluitingen
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.