Arsenic standards for ICP, TraceCERT®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
76686-100ML 72718-100ML
76686-100MLEA 112 EUR
76686-100ML 72718-100ML
Arsenic standards for ICP, TraceCERT®, Supelco®
Standaarden Kationische standaarden Arseen standaarden
Certificaten: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.