Test sieves, ASTM E 11, 200×25 mm
Leverancier: Retsch GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60.223.000020 60.223.000025 60.223.000032 60.223.000038 60.223.000045 60.223.000053 60.223.000063 60.223.000075 60.223.000090 60.223.000106 60.223.000125 60.223.000150 60.223.000180 60.223.000212 60.223.000250 60.223.000300 60.223.000355 60.223.000425 60.223.000500 60.223.000600 60.223.000710 60.223.000850 60.223.001000 60.223.001180 60.223.001400 60.223.001700 60.223.002000 60.223.002360 60.223.002800 60.223.003350 60.223.004000 60.223.004750 60.223.005600 60.223.006300 60.223.006700 60.223.008000 60.223.009500 60.223.011200 60.223.012500 60.223.013200 60.223.016000 60.223.019000 60.223.022400 60.223.025000 60.223.026500 60.223.031500 60.223.037500 60.223.045000 60.223.050000 60.223.053000 60.223.075000 60.223.090000 60.223.100000 60.223.125000
510-5721EA 912 EUR
510-5721 510-5722 510-5723 510-5724 510-5725 510-5726 510-5727 510-5728 510-5729 510-5730 510-5731 510-5732 510-5733 510-5734 510-5735 510-5736 510-5737 510-5738 510-5739 510-5740 510-5741 510-5742 510-5743 510-5744 510-5745 510-5746 510-5747 510-5748 510-5749 510-5750 510-5751 510-5752 510-5753 510-5754 510-5755 510-5756 510-5757 510-5758 510-5759 510-5760 510-5761 510-5762 510-5763 510-5764 510-5765 510-5766 510-5767 510-5768 510-5769 510-5770 510-5771 510-5772 510-5773 510-5775
Test sieves, ASTM E 11, 200×25 mm
Zeven Testzeven
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieve meshes, frames and labeling comply with standards. The sieves are tested 5 times and are delivered with test report.
Certificaten: An individual inspection certificate for test materials monitoring according to ASTM E 11 is also available on request.
Bestelinformatie: Sieves and accessories of other dimensions are available on request. Also available with perforated plate, round or square. For a full overview, please go to website.