Blood collecting system, S-Monovette® LH (liquid)
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01.1608.001 04.1939.001
SARS01.1608.001EA 0 EUR
SARS01.1608.001 SARS04.1939.001
Blood collecting system, S-Monovette® LH (liquid)
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
This blood collection system enables hygienic and easy handling in an enclosed system.
  • Minimal patient discomfort when collecting blood
  • Higher performance than Westergren method
  • HD-PE piston with PS plunger
  • With coloured paper label