Blood collecting systems, S-Monovette® CPDA1
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01.1610.001 04.1938.001
SARS01.1610.001EA 193.4 EUR
SARS01.1610.001 SARS04.1938.001
Blood collecting systems, S-Monovette® CPDA1
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
S-Monovette® is a blood collection system that combines two blood collection techniques – aspiration and vacuum. The S-Monovette® is suitable for all vein conditions and achieves an optimal sample quality, thereby producing the best results. The aspiration technique is a gentle technique for routine blood collection. Using the vacuum technique, a 'fresh' vacuum is always available.
The S-Monovette® CPDA1 is used in transfusion medicine, for blood group serology when testing for tolerance (cross-matching) in transfusions, and for cell preservation for up to 35 days, which corresponds to the preservation lifetime.
CTAD1 solution is made up of acetic acid, sodium citrate, sodium phosphate, dextrose and adenine.