Qualitix® Sterilisation Pouches
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3210.0816 3210.1326 3210.1936 3210.3040 3210.0926 3210.2537 3210.3049
SOCO3210.0816EA 14.1 EUR
SOCO3210.0816 SOCO3210.1326 SOCO3210.1936 SOCO3210.3040 SOCO3210.0926 SOCO3210.2537 SOCO3210.3049
Qualitix® Sterilisation Pouches
Zakken Sterilisatiezakken
Fitting any small to mid-size laboratory, medical, dental or veterinary device, the Qualitix® single use pouches provide easy and effective wrapping of items requiring sterilization.
  • Robust durable material
  • Quick content identification
  • Large self-sealing strip, easy to close
  • Chevron top seal for easy opening
  • Suitable for steam and EO gas sterilization
  • Two sterilization indicators
  • Five different sizes
Made with robust medical grade paper, the pouches tightly close with self-sealing strip to maintain sterility. Content remains always visible through PET/CPP transparent
laminated film.
Two chemical indicators included to confirm proper sterilisation process. Blue indicator turns dark grey after autoclaving, red indicator turns to yellow after ethylene oxide (EO) gas sterilisation.
Blue stained laminated film enables easy control of film/pouch integrity. Do not use a damaged pouch.
Certificaten: Conformity to ISO 17665-06, ISO 11135, CE.