Bottles, wide neck, with screw cap for filling and dispensing, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CE-N7Z1PI-EP CE-N7M16PI-EP CE-N7Z2PI-EP CE-N7Z4PI-EP CE-N7Z8PI-EP CEN-N7M32PI-EP
NALGCE-N7Z1PI-EPEA 0 EUR
NALGCE-N7Z1PI-EP NALGCE-N7M16PI-EP NALGCE-N7Z2PI-EP NALGCE-N7Z4PI-EP NALGCE-N7Z8PI-EP NALGCEN-N7M32PI-EP
Bottles, wide neck, with screw cap for filling and dispensing, Nalgene®
Flessen Kunststof flessen
These bottles are general purpose bottles with countless applications in the lab or field. The wide mouth makes these bottles easy to fill.
  • Excellent chemical resistance to most acids, bases and alcohols
  • Leakproof performance
  • Suitable for freezer use down to –100 °C or for shipping liquids.
Linerless caps provide the ultimate in leakproof protection without the use of liner that can wrinkle, cause leaks, or contaiminate your reagents.
Ideal for storing, shipping and packaging liquids - uniform walls provide durability and resistance to splitting or punctures.
Leveringsinformatie: Includes polypropylene screw closures.