Bumper Guards
Leverancier: ERGOMAT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HCPB100-BK HCPB100-ESD-BK HCIB120 SIBP120 LHCPB120 RCB120-ESD-BK XLSCB120 LRSB120-BK RSB120-ESD-BK
ERGOHCPB100-BKEA 37.8 EUR
ERGOHCPB100-BK ERGOHCPB100-ESD-BK ERGOHCIB120 ERGOSIBP120 ERGOLHCPB120 ERGORCB120-ESD-BK ERGOXLSCB120 ERGOLRSB120-BK ERGORSB120-ESD-BK
Bumper Guards
Meubilair Latches and Bump Protection
These Ergomat bumper guards, cart stops and edge protectors offer a variety of devices to prevent personal injury or product damage in a dynamic work environment.
  • Resistant polyurethane foam with self-adhesive attachment to rack beams, posts and uprights
  • Use anywhere workers might run into objects - supports, stands, low hanging objects
  • Round, flat, and edge surface protection
  • Yellow and black hazard striped
These guards prevent cuts and bruises from sharp edges with edge protecting bumper guards that slide over the sharp edge and holdMore Product Information
Order Now
Specificaties
Environmentally Preferable