Respiratory filters, N-series
Leverancier: HONEYWELL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1035455 1035459 1035458 1035452 1035453
DALL1035455EA 86.01 EUR
DALL1035455 DALL1035459 DALL1035458 DALL1035452 DALL1035453
Respiratory filters, N-series
Ademhalingsmaskers Herbruikbare maskers Maskerfilters
Bayonet (N-series) type filters are part of the wide range of cartridges offered to protect against many hazardous gases, vapours and/or particulates in many applications and provides high quality and a cost-effective protection.
  • Low profile and lightweight
  • Optimal control
  • Easy to attach and secure
  • Easy check up
  • Do not obstruct the field of vision and increased comfort
Useful in industries such as agriculture, automotive and part manufacturer, aviation, building and construction, chemical industries, foundry, general industry, glass industries, green spaces, industrial cleaning, iron and steel industry, laboratory, maintenance, medical and pharmaceutical, metal steel, paper industries, ship building, textile industries, utilities, water treatment, welding and wood industries.
Body material: High impact styrene, filtering material: activated and impregnated charcoal.
Certificaten: EN 14387:2004+A1:2008.