Isofreeze® racks with temperature indicator
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
95.984 95.983
SARS95.984EA 105.6 EUR
SARS95.984 SARS95.983
Isofreeze® racks with temperature indicator
Rekken Koelrekken
Isofreeze® racks are for use as a pipetting and storage station for temperature sensitive applications like enzyme analyses, PCR or cell based assays. They prevent sample damage caused by premature warming.
  • Transparent cap with white frame
  • Cooling block undergoes a clearly visible colour change from purple to pink when product leaves optimal temperature range (at 7 °C)
  • With the lid closed, the sample temperature can be maintained at ~4 °C for three hours