Screw caps for GL 18
Leverancier: QVF LABORTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HAA6011002 HAA6011102
QVFLHAA6011002EA 0 EUR
QVFLHAA6011002 QVFLHAA6011102
Screw caps for GL 18
Sluitingen Reageerbuissluitingen
Made of the most corrosion resistant, red coloured material PBT.
The cap with the bore are used as connecting cap and the cap without bore are used as sealing cap to close not engaged connections.