EDTA disodium salt solution 0.1 mol/l (0.1M) volumetric solution
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Edetic acid disodium salt dihydrate, Titriplex III, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Edetate disodium dihydrate
181670.1211 181670.1315
APPC181670.1211EA 35 EUR
APPC181670.1211 APPC181670.1315
EDTA disodium salt solution 0.1 mol/l (0.1M) volumetric solution
EDTA disodium salt
Certificaten: Traceable to NIST.
Formule: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
MDL Nummer: MFCD00150037
CAS nummer: 6381-92-6
Order Now
Specification Test Results
Titer at 20°C 0.999 - 1.001