Centrifuge tubes, disposable, plain, with screw thread finish, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP73785-15 SP73785-10 SP73785-5 SP73785-50
SCERSP73785-15EA 245 EUR
SCERSP73785-15 SCERSP73785-10 SCERSP73785-5 SCERSP73785-50
Centrifuge tubes, disposable, plain, with screw thread finish, KIMBLE®
Buisjes Centrifugebuisjes
Borosilicate glass, 33 expansion, low extractable.
  • With conical-shaped bottoms on the 5, 10, and 15 ml sizes
  • Bullet-nosed shape on the 50 ml size
  • Chemical and temperature resistance caps withstand centrifugation up to 2980×g
  • Tubes are plastic shrink-wrapped in modular trays for cleanliness and safety
Certificaten: Glass conforms USP type I and ASTM E438, Type I, Class A requirements.
Bestelinformatie: Closures to be ordered separately.