ASTM precision thermometers with safety-coating, ACCU-SAFE, Stem type
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1202005S/80 1202012S/80 1202015S/80 1202018S/80 1202022S/80 1202056S/80 1202058S/80 1202059S/80 1202062S/80 1202063S/80 1202064S/80 1202065S/80 1202066S/80 1202067S/80 1202091S/80 1202116S/80 1202117S/80 1202120S/80 1202130S/80
SCNL1202005S/80EA 0 EUR
SCNL1202005S/80 SCNL1202012S/80 SCNL1202015S/80 SCNL1202018S/80 SCNL1202022S/80 SCNL1202056S/80 SCNL1202058S/80 SCNL1202059S/80 SCNL1202062S/80 SCNL1202063S/80 SCNL1202064S/80 SCNL1202065S/80 SCNL1202066S/80 SCNL1202067S/80 SCNL1202091S/80 SCNL1202116S/80 SCNL1202117S/80 SCNL1202120S/80 SCNL1202130S/80
ASTM precision thermometers with safety-coating, ACCU-SAFE, Stem type
Thermometers Analoge thermometers
These ACCU-SAFE thermometers measures precisely and reliably without mercury from the very start.
  • Round, white back capillary form
  • High transparency for easy legibility
  • Usable as stirring thermometer
  • Suitable for calibration
The rugged encapsulation protects against splintered glass or display-fluid leakage. The protective coating is remarkable for its high transparency and quick response time allowing easily legibility. It provides temperature resistance (up to 250 °C) and hence sterilisability, and a high degree of acid and alkali resistance (hydrofluoric acid).
Leveringsinformatie: With individual serial number and certificate of conformity.
Verpakking: Each item packed in square plastic sleeve.