Handheld digital microscopes, Dino-Lite Wireless
Leverancier: Dino-Lite / IDCP B.V.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WF4515ZTL WF4915ZT WF4915ZTL WF4515ZT WF4115ZT WF4115ZTL
IDCPWF4515ZTLEA 982 EUR
IDCPWF4515ZTL IDCPWF4915ZT IDCPWF4915ZTL IDCPWF4515ZT IDCPWF4115ZT IDCPWF4115ZTL
Handheld digital microscopes, Dino-Lite Wireless
Microscopen
These microscopes from the Dino-Lite Universal series work wired via USB or wireless with the integrated WF-20 Wireless adapter.
  • Wireless and wired in one device
  • Work with different operating systems: iOS, Android or Windows
  • USB 2.0, enhanced 1,3 MP EDGE sensor
  • Integrated polariser greatly reduces glare on reflective surfaces
  • Extensive measurement functionality
  • Flexible LED control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Ideal for inspections in hard-to-reach places in factories, in the field, in workshops or in greenhouses.