1,2-Diaminocyclohexanetetraacetic acid disodium salt 0.1 M DCTA-Na4 in aqueous solution Reag. Ph. Eur. volumetrische oplossing, for complexometry, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35103-1L 35103-6X1L
35103-1LEA 180 EUR
35103-1L 35103-6X1L
1,2-Diaminocyclohexanetetraacetic acid disodium salt 0.1 M DCTA-Na4 in aqueous solution Reag. Ph. Eur. volumetrische oplossing, for complexometry, Fluka™
1,2-Diaminocyclohexanetetraacetic acid disodium salt
Formule: C₁₄H₂₀N₂Na₂O₈
CAS nummer: 5786-78-7
Order Now
Specification Test Results
Factor 0.999 - 1.001