Chemical resistant gloves, Tychem® NT430
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NT4300BU080288 NT4300BU110288
112-5478EA 0 EUR
112-5478 112-5481
Chemical resistant gloves, Tychem® NT430
Handschoenen
These 100% nitrile gloves are the perfect hybrid for jobs requiring chemical performance plus optimum fit, feel and comfort.
  • Very comfortable hybrid nitrile
  • Lightweight with 'second skin' feel
  • Embossed pattern finish provides effective, long lasting grip
Ideal for working in damp or greasy environments and for small parts handling.
Certificaten: EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016 (BACTERIA); Cat. III
Food contact approved.
Order Now
Specificaties
Model NT430
Ambidexter/handspecifiek Handspecifiek
Environmentally Preferable
Voor eenmalig gebruik/herbruikbaar Voor eenmalig gebruik
Voeringmateriaal Ongevoerd
Afwerking
Manchetstijl Gerold
EN-norm EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016 (BACTERIA)
Goedgekeurd voor voedselcontact +