High-temperature furnaces with lift up door and SiC rod, heating up to 1600 °C, LHTCT models
Leverancier: NABERTHERM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LC014H6ON1 LC014H6ON1G LC014H6ON1GT LC014H6ON1T LC034H6ON5 LC034H6ON5G LC034H6ON5GT LC034H6ON5T LC084H6ON8 LC084H6ON8G LC084H6ON8GT LC084H6ON8T
455-1854EA 6550 EUR
455-1854 455-1855 455-1856 455-1857 455-1862 455-1863 455-1864 455-1865 455-1882 455-1883 455-1884 455-1885
High-temperature furnaces with lift up door and SiC rod, heating up to 1600 °C, LHTCT models
Ovens Verassings- en moffelovens
The durability of the SiC rods in periodic use, in combination with their high heating speed, make these high-temperature furnaces good all-rounders in the laboratory. Heating times of 40 minutes to 1400 °C can be achieved, depending on the furnace model and the conditions of use.
  • SiC rod heating
  • Dual shell housing made of textured stainless steel sheets with additional fan cooling for low surface temperature
  • Only fibre materials which are not classified as carcinogenic according to TRGS 905, class 1 or 2 are used
  • Heating rods are easily replaced
  • Lift up door (LHTCT) with hot surface facing away from the operator
  • Adjustable air inlet opening, exhaust air opening in the roof
The furnace is equipped with the C550 controller with touch screen display, featuring 10 freely adjustable programs and 20 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation.
All units require 400 V three-phase power supply (400 V 3/N/PE) - only model LHTCT 01/16 is available with single phase 230 V 1/N/PE. Optional accessories e.g. over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, chimney, chimney with fan, catalytic converter, protective gas connection, manual or automatic gas supply systems are also available.
Bestelinformatie: Corresponding models with fold down doors are also available. Please search the website for more details.
Opgelet: Note: Maximum continuous operating temperature should be 50 ˚C below the max. temp. of the model shown above.
Height: Plus maximum 240 mm height for models LHTCT when open. Minutes to Tmax: If connected at 230 V 1/N/PE rsp. 400 V 3/N/PE. The working life span of the heating elements depends on several factors. Basically, increased wear must be assumed when using the furnace at high working temperatures. A typical indication of durability: 1,000 operating hours can be expected with regular use between 1500 and 1550 °C. Higher temperatures disproportionally reduce the durability of the heating elements.