Bottles, aluminium, with thread, SYSTEM PLUS® 45
Leverancier: TOURNAIRE-EMBALLAGE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4829S2 5220S1 7858S1 7418S1 4348S1 4829S1 5148S1 5376S1 5453S1 5454S1 5459S1 7419S1
215-3153EA 61 EUR
215-3153 TOUR5220S1 TOUR7858S1 TOUR7418S1 TOUR4348S1 TOUR4829S1 TOUR5148S1 TOUR5376S1 TOUR5453S1 TOUR5454S1 TOUR5459S1 TOUR7419S1
Bottles, aluminium, with thread, SYSTEM PLUS® 45
Flessen Metalen flessen
Made of pure aluminium.
  • Unbreakable, odorless
  • Very good temperature resistance
  • Leakproof with tamper-evident screw caps (must be odered separately)
  • UN-approved
Suitable for perfumes compositions, aroma chemicals, food ingredients and laboratory liquid reagents.