Chlorite ion chromatography (IC) standard, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ICS006-100ML ICS006-500ML
ICS006-100MLEA 113 EUR
ICS006-100ML ICS006-500ML
Chlorite ion chromatography (IC) standard, Supelco®
Standaarden Anionische standaarden Chloriet standaarden
Chlorite IC standard may be used in the determination of disinfection by-product, chlorite in water samples using ion chromatography technique (IC)
Certificaten: his certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.