ESD safety ankle boots, lace up, Wellmaxx, Sensation XX10 mid
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
769901/36 769901/37 769901/38 769901/39 769901/40 769901/41 769901/42 769901/43 769901/44 769901/45 769901/46 769901/47 769901/48
114-8843EA 94.9 EUR
114-8843 114-8844 114-8845 114-8846 114-8847 114-8848 114-8849 114-8850 114-8851 114-8852 114-8853 114-8854 114-8855
ESD safety ankle boots, lace up, Wellmaxx, Sensation XX10 mid
Schoenen en laarzen
ESD safety ankle boots in denim look made from hydrophobised textile material and hydrophobised nubuck leather, a padded tongue and a PU/PU sole WELLMAXX L10.
  • Bioactive fabric lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), grey
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
  • Sole core made of Infinergy® from BASF
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/grey
Certificaten: EN 61340-5-1 (ESD), EN ISO 20345; S3, SRC, form B