Fluka™ Boiling stones ('Anti-Bumping'), granules 1-3 mm
Leverancier: Honeywell Research Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85106-500GEA 147 EUR
85106-500G
Fluka™ Boiling stones ('Anti-Bumping'), granules 1-3 mm
Kookstenen
  • Sieve analysis > 1 mm: min 95%