ESD safety shoes, lace-up, Impulse EA blue low, S2
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7272601/40 7272601/41 7272601/42 7272601/43 7272601/44 7272601/45 7272601/46 7272601/47 7272601/48 7272602/40 7272602/41 7272602/42 7272602/43 7272602/44 7272602/45 7272602/46 7272602/47 7272602/48 7272603/40 7272603/41 7272603/42 7272603/43 7272603/44 7272603/45 7272603/46 7272603/47 7272603/48
ELTN7272601/40EA 88.9 EUR
ELTN7272601/40 ELTN7272601/41 ELTN7272601/42 ELTN7272601/43 ELTN7272601/44 ELTN7272601/45 ELTN7272601/46 ELTN7272601/47 ELTN7272601/48 ELTN7272602/40 ELTN7272602/41 ELTN7272602/42 ELTN7272602/43 ELTN7272602/44 ELTN7272602/45 ELTN7272602/46 ELTN7272602/47 ELTN7272602/48 ELTN7272603/40 ELTN7272603/41 ELTN7272603/42 ELTN7272603/43 ELTN7272603/44 ELTN7272603/45 ELTN7272603/46 ELTN7272603/47 ELTN7272603/48
ESD safety shoes, lace-up, Impulse EA blue low, S2
Schoenen en laarzen
Leather-free, sporty and lightweight ESD safety shoes made from microfibre, reflective materials, a full padded bellows tongue and a slip resistant dual-density TPU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Replaceable insole; orthopaedic insoles possible
  • Full-length semi-orthopaedic inlay sole ESD black
  • Soft fleece ESD insole
  • Elastic lacing system with individual quick locking system
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Blue/black
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC, form A