Molecular sieve deperoxidation, beads 10-20 mesh for the deperoxidation and drying of solvents, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
69805-1KG 69805-250G
69805-1KGEA 600 EUR
69805-1KG 69805-250G
Molecular sieve deperoxidation, beads 10-20 mesh for the deperoxidation and drying of solvents, Fluka™
Molecular sieve deperoxidation
Order Now
Specification Test Results
Appearance complying
Loss on ignition (900°C) Max. 3 %
Grain size 0.85-2.0 mm Min. 95 %
Capacity ≥20 % absorption capacity (water)