ESD safety shoes, lace-up, William Low
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7273101/40 7273101/41 7273101/42 7273101/43 7273101/44 7273101/45 7273101/46 7273101/47 7273101/48 7273102/40 7273102/41 7273102/42 7273102/43 7273102/44 7273102/45 7273102/46 7273102/47 7273102/48 7273103/40 7273103/41 7273103/42 7273103/43 7273103/44 7273103/45 7273103/46 7273103/47 7273103/48 7274101/40 7274101/41 7274101/42 7274101/43 7274101/44 7274101/45 7274101/46 7274101/47 7274101/48 7274102/40 7274102/41 7274102/42 7274102/43 7274102/44 7274102/45 7274102/46 7274102/47 7274102/48 7274103/40 7274103/41 7274103/42 7274103/43 7274103/44 7274103/45 7274103/46 7274103/47 7274103/48
ELTN7273101/40EA 85.9 EUR
ELTN7273101/40 ELTN7273101/41 ELTN7273101/42 ELTN7273101/43 ELTN7273101/44 ELTN7273101/45 ELTN7273101/46 ELTN7273101/47 ELTN7273101/48 ELTN7273102/40 ELTN7273102/41 ELTN7273102/42 ELTN7273102/43 ELTN7273102/44 ELTN7273102/45 ELTN7273102/46 ELTN7273102/47 ELTN7273102/48 ELTN7273103/40 ELTN7273103/41 ELTN7273103/42 ELTN7273103/43 ELTN7273103/44 ELTN7273103/45 ELTN7273103/46 ELTN7273103/47 ELTN7273103/48 ELTN7274101/40 ELTN7274101/41 ELTN7274101/42 ELTN7274101/43 ELTN7274101/44 ELTN7274101/45 ELTN7274101/46 ELTN7274101/47 ELTN7274101/48 ELTN7274102/40 ELTN7274102/41 ELTN7274102/42 ELTN7274102/43 ELTN7274102/44 ELTN7274102/45 ELTN7274102/46 ELTN7274102/47 ELTN7274102/48 ELTN7274103/40 ELTN7274103/41 ELTN7274103/42 ELTN7274103/43 ELTN7274103/44 ELTN7274103/45 ELTN7274103/46 ELTN7274103/47 ELTN7274103/48
ESD safety shoes, lace-up, William Low
Schoenen en laarzen
Leather-free, ESD safety shoe made from microfibre/mesh, reflective materials, a padded tongue and a slip resistant dual-density TPU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Full-length semi-orthopaedic inlay sole ESD black
  • Anatomically shaped insole and ankle support for improved stability
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in dry areas where there is a risk of penetration from pointed and sharp objects.
Colour: Black/blue or black/red
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC, form A.