ESD safety shoes, lace-up, Anthony Low
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7275103/40 7275103/41 7275103/42 7275103/43 7275103/44 7275103/45 7275103/46 7275103/47 7275103/48 1727511/36 1727511/37 1727511/38 1727511/39 1727512/36 1727512/37 1727512/38 1727512/39 1727513/36 1727513/37 1727513/38 1727513/39
ELTN7275103/40EA 87.9 EUR
ELTN7275103/40 ELTN7275103/41 ELTN7275103/42 ELTN7275103/43 ELTN7275103/44 ELTN7275103/45 ELTN7275103/46 ELTN7275103/47 ELTN7275103/48 ELTN1727511/36 ELTN1727511/37 ELTN1727511/38 ELTN1727511/39 ELTN1727512/36 ELTN1727512/37 ELTN1727512/38 ELTN1727512/39 ELTN1727513/36 ELTN1727513/37 ELTN1727513/38 ELTN1727513/39
ESD safety shoes, lace-up, Anthony Low
Schoenen en laarzen
Leather-free, sporty and lightweight ESD safety shoe made from microfibre, a full padded bellows tongue and a slip resistant dual-density TPU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Full-length semi-orthopaedic inlay sole ESD black
  • Anatomically shaped insole and ankle support for improved stability
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in dry areas and where there is a risk of penetration from pointed and sharp objects. The sizes 36 to 39 are ideally tailored to the ergonomics of female feet.
Colour: Black/red
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC, form A