Multiport connector caps, GL 32, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
292611902 292621903 292601901
SCOT292611902EA 75.5 EUR
SCOT292611902 SCOT292621903 SCOT292601901
Multiport connector caps, GL 32, DURAN®
Sluitingen Flesdoppen
The reusable GL 32 caps allow the smaller 50 ml round or 100 ml DURAN® square bottles to be used be used for sampling, liquid feeding or transfer. Flexible options of 2, 3, or 4 stainless steel tubing connectors.The barbed male tubing connector are suitable for tubing with a 3,2 mm inner diameter such as BPT peristaltic pump tubing.
  • Autoclavable (121 °C / 15 minutes)
  • Temperature resistance up to 180 °C
  • Reusable
  • All components are chemically resistant
Ideal for small-scale bioprocess applications such as media supply for small bioreactors, supplyingMore Product Information