ESD safety ankle boots, lace-up, Matthew TPU Mid
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7676101/40 7676101/41 7676101/42 7676101/43 7676101/44 7676101/45 7676101/46 7676101/47 7676101/48 7676102/40 7676102/41 7676102/42 7676102/43 7676102/44 7676102/45 7676102/46 7676102/47 7676102/48 7676103/40 7676103/41 7676103/42 7676103/43 7676103/44 7676103/45 7676103/46 7676103/47 7676103/48 1767601/36 1767601/37 1767601/38 1767601/39 1767602/36 1767602/37 1767602/38 1767602/39 1767603/36 1767603/37 1767603/38 1767603/39
ELTN7676101/40EA 91.9 EUR
ELTN7676101/40 ELTN7676101/41 ELTN7676101/42 ELTN7676101/43 ELTN7676101/44 ELTN7676101/45 ELTN7676101/46 ELTN7676101/47 ELTN7676101/48 ELTN7676102/40 ELTN7676102/41 ELTN7676102/42 ELTN7676102/43 ELTN7676102/44 ELTN7676102/45 ELTN7676102/46 ELTN7676102/47 ELTN7676102/48 ELTN7676103/40 ELTN7676103/41 ELTN7676103/42 ELTN7676103/43 ELTN7676103/44 ELTN7676103/45 ELTN7676103/46 ELTN7676103/47 ELTN7676103/48 ELTN1767601/36 ELTN1767601/37 ELTN1767601/38 ELTN1767601/39 ELTN1767602/36 ELTN1767602/37 ELTN1767602/38 ELTN1767602/39 ELTN1767603/36 ELTN1767603/37 ELTN1767603/38 ELTN1767603/39
ESD safety ankle boots, lace-up, Matthew TPU Mid
Schoenen en laarzen
Leather-free ESD safety ankle boots made from hydrophobised microfibre and hydrophobised textile materials, reflective materials, a fully padded bellows tongue and a slip resistant dual density TPU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Replaceable insole; orthopaedic insoles possible
  • Semi-orthopaedic inlay sole ESD black
  • Anatomically-shaped insole and ankle support for improved stability
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects. The sizes 36 to 39 are ideally tailored to the ergonomics of female feet.
Colour: Black/red
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, form B