Temperature Probes, PT 100, Lemo®, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P1762-10 P1762-15 P1762-20 P1762-25 P1762-30
BOHLP1762-10EA 330 EUR
BOHLP1762-10 BOHLP1762-15 BOHLP1762-20 BOHLP1762-25 BOHLP1762-30
Temperature Probes, PT 100, Lemo®, BOLA
Temperatuursondes
Thermocouple in a PTFE-encapsulated stainless steel tube.
  • Universal chemical resistance
  • Temperature resistance from –200 to +250 °C
  • Temperature probe of tip Ø 6 mm and collar ring Ø 12 mm
  • With white PTFE-coated cable (length of 1,5 m) and mounted Lemo® plug size 1,4-wire system
  • Typical response times of T 50 is 7 to 12 seconds and T 90 is 14 to 16 seconds
Used for temperature measurement in aggressive liquids and cable provides flexible connection from measuring device to medium.
Certificaten: Conforms to FDA