VWR®, Water bath stabiliser, AQUA RESIST WB
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86174.360EA 86.3 EUR
86174.360 86174.440
VWR®, Water bath stabiliser, AQUA RESIST WB
Reinigingsmiddelen Reagentia voor bescherming van waterbaden
A ready to use, liquid product designed to stabilise and protect water baths, cooling circuits, water circuits and incubators.
  • Working temperature up to 70 °C
  • Non corrosive for conventional materials
  • Good wetting characteristics
  • Low foaming
  • Blue dye for visualisation
AQUA RESIST WB is characterised by a specific combination of several ingredients. Surfactants cause stabilisation of the water with simultaneous good wetting. Stabilisers and corrosion inhibitors ensure high material compatibility and thus for a wide range of applications even with longer exposure times. The blue colour makes visual inspection of the existing product possible.