ESD safety shoes, lace-up, uvex 2 xenova®, 9505
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9505.7/38 9505.7/39 9505.7/40 9505.7/41 9505.7/42 9505.7/43 9505.7/44 9505.7/45 9505.7/46 9505.7/47 9505.7/48 9505.7/49 9505.7/50 9505.7/51 9505.7/52 9505.8/38 9505.8/39 9505.8/40 9505.8/41 9505.8/42 9505.8/43 9505.8/45 9505.8/46 9505.8/47 9505.8/48 9505.8/49 9505.8/50 9505.8/51 9505.8/52
UVEX9505.7/38EA 86.7 EUR
UVEX9505.7/38 UVEX9505.7/39 UVEX9505.7/40 UVEX9505.7/41 UVEX9505.7/42 UVEX9505.7/43 UVEX9505.7/44 UVEX9505.7/45 UVEX9505.7/46 UVEX9505.7/47 UVEX9505.7/48 UVEX9505.7/49 UVEX9505.7/50 UVEX9505.7/51 UVEX9505.7/52 UVEX9505.8/38 UVEX9505.8/39 UVEX9505.8/40 UVEX9505.8/41 UVEX9505.8/42 UVEX9505.8/43 UVEX9505.8/45 UVEX9505.8/46 UVEX9505.8/47 UVEX9505.8/48 UVEX9505.8/49 UVEX9505.8/50 UVEX9505.8/51 UVEX9505.8/52
ESD safety shoes, lace-up, uvex 2 xenova®, 9505
Schoenen en laarzen
Very lightweight fashionable uvex 2 xenova® perforated S1 safety shoes made from synthetic materials, suitable for people allergic to chrome. All sole materials are free of silicones, plasticisers and other paint-wetting impairment substances.
  • Compact design, anatomical shape, good lateral stability, no thermal conductivity
  • Climazone technology for improved comfort
  • Breathable materials as well as the perforated upper material
  • Removable anti-static comfortable insole, with moisture transport system
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Shock absorption around the heel and forefoot
  • Metal-free uvex xenova® toe cap
  • Fulfils the ESD requirement: Discharge resistance <35 megohm
Used for medium-duty applications.
It has soft padded collar for wearer comfort.
Colour: Black/red
Certificaten: EN 61340 (ESD); EN ISO 20345:2011; S1, SRC