Spoon spatulas, micro, powder, stainless steel
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
625.150 627.151 627.186 625.185 625.210 627.211
RSGA625.150EA 35 EUR
RSGA625.150 RSGA627.151 RSGA627.186 RSGA625.185 RSGA625.210 RSGA627.211
Spoon spatulas, micro, powder, stainless steel
Spatels
These are made of stainless steel 18/10.
  • Very good chemical resistance
  • Very good thermal resistance
  • Quick and easy to clean
With a micro spoon at one end and a micro spatula at the other end.