Titrimetric rapid tests, alkalinity, VISOCOLOR®
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
915207 915007
MANA915207EA 41.1 EUR
MANA915207 MANA915007
Titrimetric rapid tests, alkalinity, VISOCOLOR®
Milieutestkits
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
All compounds which cause a pH increase above pH 7 are determined, e.g. hydroixde,carbonate, hydrogencarbonate etc…
Note: to differentiate between hydoxide, carbonate and hydrogen carbonate  you should use the test kit VISOCOLOR® HE Carbonate hardness C 20 (see German Standard Methods DIN 38 409-H7).