Titrimetric rapid tests, sulfite, VISOCOLOR®
Leverancier: MACHEREY-NAGEL
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
915208
931095
915008
MANA915208EA
67
EUR
MANA915208
MANA931095
MANA915008
Titrimetric rapid tests, sulfite, VISOCOLOR®
Milieutestkits
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
- Portable
- Environmentally friendly
- Convenient handling
- Application-oriented measuring ranges
Sulphite ions are not present in natural unpolluted waters, however they often appear in large quantities in industrial waste waters (e. g. paper mills, dye works).