ESD safety ankle boots, lace-up, uvex 2, Xenova®, 9566
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9556.1/38 9556.1/39 9556.1/40 9556.1/41 9556.1/42 9556.1/43 9556.1/44 9556.1/45 9556.1/46 9556.1/47 9556.1/48 9556.1/49 9556.1/50 9556.1/51 9556.1/52 9556.2/38 9556.2/39 9556.2/40 9556.2/41 9556.2/42 9556.2/43 9556.2/44 9556.2/45 9556.2/46 9556.2/47 9556.2/48 9556.2/49 9556.2/50 9556.2/51 9556.2/52 9556.3/38 9556.3/39 9556.3/40 9556.3/41 9556.3/42 9556.3/43 9556.3/44 9556.3/45 9556.3/46 9556.3/47 9556.3/48 9556.3/49 9556.3/50 9556.3/51 9556.3/52
UVEX9556.1/38EA 109 EUR
UVEX9556.1/38 UVEX9556.1/39 UVEX9556.1/40 UVEX9556.1/41 UVEX9556.1/42 UVEX9556.1/43 UVEX9556.1/44 UVEX9556.1/45 UVEX9556.1/46 UVEX9556.1/47 UVEX9556.1/48 UVEX9556.1/49 UVEX9556.1/50 UVEX9556.1/51 UVEX9556.1/52 UVEX9556.2/38 UVEX9556.2/39 UVEX9556.2/40 UVEX9556.2/41 UVEX9556.2/42 UVEX9556.2/43 UVEX9556.2/44 UVEX9556.2/45 UVEX9556.2/46 UVEX9556.2/47 UVEX9556.2/48 UVEX9556.2/49 UVEX9556.2/50 UVEX9556.2/51 UVEX9556.2/52 UVEX9556.3/38 UVEX9556.3/39 UVEX9556.3/40 UVEX9556.3/41 UVEX9556.3/42 UVEX9556.3/43 UVEX9556.3/44 UVEX9556.3/45 UVEX9556.3/46 UVEX9556.3/47 UVEX9556.3/48 UVEX9556.3/49 UVEX9556.3/50 UVEX9556.3/51 UVEX9556.3/52
ESD safety ankle boots, lace-up, uvex 2, Xenova®, 9566
Schoenen en laarzen
Metal-free, lightweight, flexible and robust safety shoe made from water-repellent, full-grain smooth leather upper material, soft padded tongue and collar, and a dual-density slip-resistant Xenova® PU sole resistant to heat, chemicals, oils and gasoline.
  • Compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
  • Anti-twist rear cap provides more stability and kink protection
  • Climazone technology for improved comfort
  • Non-marking, with excellent cushioning and abrasion-resistant outsole
  • Anti-static comfort insole with moisture control system; orthopaedic insoles
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Shock absorbing
  • Fulfils the ESD requirement: Discharge resistance <35 megohm
  • PU-coated leather scuffcap, non-metallic, penetration-resistant midsole
Ideal for applications such as mechanical and plant engineering, metal processing, energy (generation, supply and grid operation), municipal operations, motorway maintenance, railways, transport & logistics, airports, trades, ancillary construction work, horticulture and landscaping.
Colour: Black/blue
Certificaten: EN 61340 (ESD); EN ISO 20345: 2011 S3 SRC