Attached Lid Containers
Leverancier: MULTIROIR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BA12145 BA12146 BA12147 BA12148
216-2248EA 46.5 EUR
216-2248 216-2249 216-2251 216-2252
Attached Lid Containers
Dozen Transportdozen
These containers, with robust attached lids, are designed to stack when full and nest when empty. The recesses of the hinged lid ensure high stability.
  • Made of of heavy-duty polypropylene, rigid, robust and practical
  • Containers nest for maximum space usage with lids open
  • Blue
  • When empty, saves up to 75% valuable storage space
  • Easy to clean interiors
The attached lid containers can be secured with plastic seals and transported with trolleys.