Basic Violet 3 solution
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: CI 42555
911517ZAEA 62.7 EUR
911517ZA 911517ZAMP
Basic Violet 3 solution
Basic Violet 3
Primary stain for Gram staining
Formule: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Nummer: MFCD00011750
CAS nummer: 548-62-9
Merck Index: 13,04406