Poly(methacrylic acid sodium salt) analytical standards, for GPC, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
02351-250MG 02356-250MG 02363-250MG
02351-250MGEA 317 EUR
02351-250MG 02356-250MG 02363-250MG
Poly(methacrylic acid sodium salt) analytical standards, for GPC, Supelco®
Standaarden Standaarden voor chromatografie
Certificaten: Exact values can be taken from the accompanying certificate of analysis
Opgelet: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1633c32ebafa777837a6eefbe6c02e1eff6ee7b7&vid=d09cc7e22eb679bfa432ec666357b4394a46d002&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=799528a4f18855b0f49fe2d32b39b9bc470b61c8&vid=799528a4f18855b0f49fe2d32b39b9bc470b61c8&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86973b6fdc08a9dca6bf11fb4daae2243bee68a4&vid=86973b6fdc08a9dca6bf11fb4daae2243bee68a4&prid=web&clid=SAPDAM