Erlenmeyer shake flasks, with DuoCap, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WPFPC0125S WPFPC2000S WPFPC1000S WPFBC2000S WPFPC0500S WPFPC3000S WPFBC0250S WPFBC0500S WPFBC1000S
WHEAWPFPC0125SEA 549 EUR
WHEAWPFPC0125S WHEAWPFPC2000S WHEAWPFPC1000S WHEAWPFBC2000S WHEAWPFPC0500S WHEAWPFPC3000S WHEAWPFBC0250S WHEAWPFBC0500S WHEAWPFBC1000S
Erlenmeyer shake flasks, with DuoCap, WHEATON®
Kolven Erlenmeyers
These Erlenmeyer flasks are made of lexan polycarbonate resin that has outstanding mechanical, optical, electrical and thermal properties.
  • Manufactured in a class 10000 cleanroom
  • DuoCAP® allows the option of sterile air exchange or a leak resistant seal
  • Certified pyrogen-, DNase- and RNase-free
  • Gamma irradiated, sterile to SAL 10-6
  • Re-autoclavable and reusable
  • Supports both aerobic and anaerobic culture
The resin conforms to flame retardant standards, is scratch resistant and retains clarity after gamma irradiation. It also meets the requirements of United States Pharmacopeia (USP) XXII Class VI.
Flasks fit all standard shaking incubator systems and can be placed on any platform.
Ideal for all suspension cell cultures, including baculovirus, microbial and algae cultures, as well as media preparation, storage and all related applications.
Leveringsinformatie: Both flasks are supplied standard with a patented vented DuoCap.
Verpakking: Packaged particulate free and wrapped in an easy tear, sterile bag.