Sand (sea sand), AnalaR NORMAPUR® zA, purified_acid, calcinated
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86152.290EA 55.7 EUR
86152.290 86152.360 86152.460
Sand (sea sand), AnalaR NORMAPUR® zA, purified_acid, calcinated
Sand (sea sand)
Formule: SiO₂
MW: 60.08 g/mol
Kookpunt: 2230
Smeltpunt: 1710
CAS nummer: 14808-60-7
Order Now
Specification Test Results
Particle size (0.1-0.3 mm) Min. 80 %
Iron soluble in acid Max. 100 ppm
Loss on ignition (900°C) Max. 0.1 %
Substances soluble in HCl Max. 0.2 %
Cl (Chloride) Max. 50 ppm