Biological indicators, spore suspensions
Leverancier: LIOFILCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
82003 82016 82009 82012 82031 82017 82010 82004 82014 82030
82003.EA 999 EUR
82003. 82016. 82009. 82012. 82031. 82017. 82010. 82004. 82014. 82030.
Biological indicators, spore suspensions
Sterilisatie-indicatoren
Biological indicators for validating the sterilisation using steam (autoclaves), VHP (isolators), dry heat and irradiation processes.
Order Now
Specificaties
Environmentally Preferable