Biological indicators, strips
Leverancier: LIOFILCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
91055 91067 91155 91072 91071 91064
LIOF91055EA 221 EUR
LIOF91055 91067. 91155. 91072. 91071. 91064.
Biological indicators, strips
Sterilisatie-indicatoren
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).