Colorimetric Rapid Tests, Aluminium, VISOCOLOR®
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
931006 931206
MANA931006EA 85.2 EUR
MANA931006 MANA931206
Colorimetric Rapid Tests, Aluminium, VISOCOLOR®
Milieutestkits
Colorimetric determination of aluminium using VISOCOLOR® ECO Aluminium
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
Aluminium is the most common metal in our anthroposphere and after oxygen and silica even the most common element of the earth’s crust. Because of its big affinity to oxygen, aluminium does not exist in elemental form in nature, but in different oxidized compounds.
In swimming pools, aluminium salts are used as flocculant for the water treatment. For drinking water, the WHO recommends a guide value of 0.2 mg/l Al3+. In accordance to the EU council guideline 98/83/EEC, the threshold value for drinking water is 0.2 mg/l Al3+. In natural water, the concentration of aluminium compounds is even low, but in waste water aluminium can appear in higher concentrations, e.g. at electroplating companies or paper mills. Different national regulations tolerate 2 to 3 mg/l Al3+ in effluents from various industries (metal, electroplating and printing industries).
Order Now
Specificaties
Environmentally Preferable