Colorimetric rapid tests, sulphide, VISOCOLOR® ECO
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
931094 931294
MANA931094EA 79.4 EUR
MANA931094 MANA931294
Colorimetric rapid tests, sulphide, VISOCOLOR® ECO
Milieutestkits
Colorimetric determination of sulphide by DPD method.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
In water sulphides can be present as dissolved hydrogen sulphide or as hydrosulphide or sulphide ions.
Suitable for sea water testing.