Industrial gloves, HyFlex® 11-849
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-849/6 11-849/8 11-849/9 11-849/10 11-849/11 11-849/7
ANSE11-849/6EA 85.9 EUR
ANSE11-849/6 ANSE11-849/8 ANSE11-849/9 ANSE11-849/10 ANSE11-849/11 ANSE11-849/7
Industrial gloves, HyFlex® 11-849
Handschoenen
The fully dipped design offers 360° protection against scratches, snags, and dirt, while the thin foam nitrile coating allow the hand to breath for better comfort.
  • 360° breathable coating provides additional protection
  • Ansell GRIP™ technology for a sure grip
  • Ergonomic design for a secured and comfortable fit
Designed to provide all around comfort and protection across scratches, snags, and dirt. The 360° breathable coating provides additional protection on the back hand, without compromising on comfort. FORTIX™ all around protection ensures a longer continuous wear time in abrasive dry environments. No transfer of silicone contaminants to metal parts prior to painting.
Certificaten: EN 388:2016 (4121A); Cat. II
Verpakking: Gloves are packaged 12 pairs per bag, 12 bags per carton.