Millex® Syringe Filters, HPF, LCR (hydrophilic PTFE) with Glass Fibre Pre-filter
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SLCRM25NK SLCRM25NS SLLGM25NK SLLGM25NS
MILFSLCRM25NKEA 3170 EUR
MILFSLCRM25NK MILFSLCRM25NS MILFSLLGM25NK MILFSLLGM25NS
Millex® Syringe Filters, HPF, LCR (hydrophilic PTFE) with Glass Fibre Pre-filter
Filters Spuitfilters
These filters include multiple media: a graduated prefilter and a membrane filter for high throughput filtration of viscous or particle-laden solutions.
  • High-density polyethylene housing compatible with the most commonly used solvents
  • Multi-layer filter can improve throughput of high particulate/viscous samples
Applications include analytical sample preparation, wine analysis and general particulate removal.